3-Cyclohexen-1-carbonsäure 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Cyclohex-3-enecarboxylic acid, 3-Cyclohexenecarboxylic acid, 1,2,3,6-Tetrahydrobenzoic Acid

OR30377-100G OR30377-500G OR30377-25G
APOSOR30377-100GEA 34.8 EUR
APOSOR30377-100G APOSOR30377-500G APOSOR30377-25G
3-Cyclohexen-1-carbonsäure 98%
3-Cyclohexen-1-carbonsäure
Formel: C₇H₁₀O₂
Molecular Weight: 126.16 g/mol
Schmelzpunkt: 17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013781
CAS-Nummer: 4771-80-6
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR