Ethyl-3-chlorpicolinat
Lieferant: Apollo Scientific
|
Achtung
|
OR303770-1G
OR303770-500MG
APOSOR303770-1GEA
0
EUR
APOSOR303770-1G
APOSOR303770-500MG
Ethyl-3-chlorpicolinat
Ethyl-3-chlorpicolinat
Formel:
C₈H₈ClNO₂ Siedepunkt: 104…106 °C/0,7mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD11656307 CAS-Nummer: 128073-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...