6-[(4-Bromophenyl)sulphanyl]-3-pyridinylamine
Lieferant: Apollo Scientific
OR303788-500MG
APOSOR303788-500MGEA
336
EUR
APOSOR303788-500MG
6-[(4-Bromophenyl)sulphanyl]-3-pyridinylamine
6-[(4-Bromophenyl)sulphanyl]-3-pyridinylamine
Formel:
C₁₁H₉BrN₂S Molecular Weight: 281,18 g/mol Schmelzpunkt: 81…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01927961 CAS-Nummer: 353277-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...