6-(Methylsulphonyl)-3-pyridinamine

Lieferant: Apollo Scientific

OR303789-500MG
APOSOR303789-500MGEA 0 EUR
APOSOR303789-500MG
6-(Methylsulphonyl)-3-pyridinamine
6-(Methylsulphonyl)-3-pyridinamine
Formel: C₆H₈N₂O₂S
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09941635
CAS-Nummer: 187143-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR