Methyl-4-aminopicolinat 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Aminopyridine-2-carboxylic acid methyll ester, Methyl 4-Aminopyridine-2-carboxylate, 4-Aminopicolinic acid methyll ester, Methyl 4-Aminopicolinate
OR303823-1G
OR303823-5G
OR303823-25G
APOSOR303823-1GEA
71.2
EUR
APOSOR303823-1G
APOSOR303823-5G
APOSOR303823-25G
Methyl-4-aminopicolinat 97%
Methyl-4-aminopicolinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.1500 g/mol Schmelzpunkt: 129…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00956000 CAS-Nummer: 71469-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...