Methyl-4-aminopicolinat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Aminopyridine-2-carboxylic acid methyll ester, Methyl 4-Aminopyridine-2-carboxylate, 4-Aminopicolinic acid methyll ester, Methyl 4-Aminopicolinate

OR303823-1G OR303823-5G OR303823-25G
APOSOR303823-1GEA 71.2 EUR
APOSOR303823-1G APOSOR303823-5G APOSOR303823-25G
Methyl-4-aminopicolinat 97%
Methyl-4-aminopicolinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.1500 g/mol
Schmelzpunkt: 129…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00956000
CAS-Nummer: 71469-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR