4,6-Dichlornicotinamid

Lieferant: Apollo Scientific

OR303826-1G
APOSOR303826-1GEA 0 EUR
APOSOR303826-1G
4,6-Dichlornicotinamid
4,6-Dichlornicotinamid
Formel: C₆H₄Cl₂N₂O
Molecular Weight: 191,0200 g/mol
Schmelzpunkt: 153…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646073
CAS-Nummer: 70593-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR