2,4-Dichlor-3-formylpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Dichloronicotinaldehyde, 2,4-Dichloropyridine-3-carboxaldehyde, 2,4-Dichloro-3-formylpyridine, 2,4-Dichloro-3-pyridinecarboxaldehyde
OR303843-25G
APOSOR303843-25GEA
149
EUR
APOSOR303843-25G
2,4-Dichlor-3-formylpyridin 98%
2,4-Dichlor-3-formylpyridin
Formel:
C₆H₃Cl₂NO Molecular Weight: 176.00 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437909 CAS-Nummer: 134031-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...