2-Chloro-4-(chloromethyl)-6-methoxypyridine 95%

Lieferant: Apollo Scientific

OR303859-1G OR303859-500MG
APOSOR303859-1GEA 894 EUR
APOSOR303859-1G APOSOR303859-500MG
2-Chloro-4-(chloromethyl)-6-methoxypyridine 95%
2-Chloro-4-(chloromethyl)-6-methoxypyridine
Formel: C₇H₇Cl₂NO
Molecular Weight: 192.05 g/mol
Schmelzpunkt: 40…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12405630
CAS-Nummer: 727648-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR