6-[(4-Bromophenyl)sulphonyl]-3-pyridinylamine

Lieferant: Apollo Scientific

OR303877-500MG
APOSOR303877-500MGEA 560 EUR
APOSOR303877-500MG
6-[(4-Bromophenyl)sulphonyl]-3-pyridinylamine
6-[(4-Bromophenyl)sulphonyl]-3-pyridinylamine
Formel: C₁₁H₉BrN₂O₂S
Molecular Weight: 313,18 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16140339
CAS-Nummer: 1221791-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR