2,5-Dichlor-4-formylpyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dichloroisonicotinaldehyde
OR303891-5G
APOSOR303891-5GEA
97.3
EUR
APOSOR303891-5G
2,5-Dichlor-4-formylpyridin 97%
2,5-Dichlor-4-formylpyridin
Formel:
C₆H₃Cl₂NO Schmelzpunkt: 73…74 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06410679 CAS-Nummer: 102645-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...