Diethyl-3-cyano-6-cyclopropyl-2,5-pyridinedicarboxylate

Lieferant: Apollo Scientific

OR303898-500MG
APOSOR303898-500MGEA 661 EUR
APOSOR303898-500MG
Diethyl-3-cyano-6-cyclopropyl-2,5-pyridinedicarboxylate
Diethyl-3-cyano-6-cyclopropyl-2,5-pyridinedicarboxylate
Formel: C₁₅H₁₆N₂O₄
Molecular Weight: 288,31 g/mol
Schmelzpunkt: 117…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16140279
CAS-Nummer: 1221792-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR