Ethyl 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate

Lieferant: Apollo Scientific

OR303911-5G
APOSOR303911-5GEA 275 EUR
APOSOR303911-5G
Ethyl 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Formel: C₁₀H₁₂N₄O₂
Molecular Weight: 220.228 g/mol
Schmelzpunkt: 192…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03069208
CAS-Nummer: 2627-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR