4-Carbamoylphenylboronsäurepinakolester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Aminocarbonylphenylboronic acid, pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide, 4-(Aminocarbonyl)phenylboronic acid pinacol ester

OR303954-500MG
APOSOR303954-500MGEA 21.9 EUR
APOSOR303954-500MG
4-Carbamoylphenylboronsäurepinakolester 98%
4-Carbamoylphenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₃
Molecular Weight: 247,0979 g/mol
Schmelzpunkt: 175…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663887
CAS-Nummer: 179117-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR