5-Ethyl-2-methylpiperidine

Lieferant: Apollo Scientific
Achtung

OR30399-1G OR30399-5G
APOSOR30399-1GEA 0 EUR
APOSOR30399-1G APOSOR30399-5G
5-Ethyl-2-methylpiperidine
5-Ethyl-2-methylpiperidine
Formel: C₈H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD00023134
CAS-Nummer: 104-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR