2-(Benzylamino)-2-methylpropanenitrile

Lieferant: Apollo Scientific

OR303996-500MG
APOSOR303996-500MGEA 214 EUR
APOSOR303996-500MG
2-(Benzylamino)-2-methylpropanenitrile
2-(Benzylamino)-2-methylpropanenitrile
Formel: C₁₁H₁₄N₂
Molecular Weight: 174.242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10014472
CAS-Nummer: 99840-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR