(2R)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%
Lieferant: Apollo Scientific
OR304009-5G
APOSOR304009-5GEA
156
EUR
APOSOR304009-5G
(2R)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol
Formel:
C₁₇H₂₁NO Molecular Weight: 255,3600 g/mol Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318247 CAS-Nummer: 86695-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...