(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 97+%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Amino-3-methyl-1,1-diphenylbutan-1-ol

OR304010-5G
APOSOR304010-5GEA 94.4 EUR
APOSOR304010-5G
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 97+%
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol

Used in enantioselective reduction of ketones and oxime ethers. J.Chem. Soc. Perkin Trans: .I, 1985, 2039

Formel: C₁₇H₂₁NO
Molecular Weight: 255.36 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318248
CAS-Nummer: 78603-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR