Iminodibenzyl
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
10,11-Dihydro-5H-dibenzo[b,f]azepine
OR30408-5G
APOSOR30408-5GEA
21.9
EUR
APOSOR30408-5G
Iminodibenzyl
Iminodibenzyl
Formel:
C₁₄H₁₃N Molecular Weight: 195,27 g/mol Schmelzpunkt: 105…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005070 CAS-Nummer: 494-19-9 EINECS: 207-787-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...