6-Methyl-3,4-dihydro-2H-pyran-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dihydro-6-methyl-2H-pyran-5-carboxylic acid

OR30416-250MG
APOSOR30416-250MGEA 190 EUR
APOSOR30416-250MG
6-Methyl-3,4-dihydro-2H-pyran-5-carboxylic acid
6-Methyl-3,4-dihydro-2H-pyran-5-carboxylic acid
Formel: C₇H₁₀O₃
Molecular Weight: 142,16 g/mol
Schmelzpunkt: 119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179417
CAS-Nummer: 5399-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR