5-Hydroxy-o-toluylsäure 98%

Lieferant: Apollo Scientific

Synonyme: 5-Hydroxy-2-methylbenzoic acid

OR304202-25G
APOSOR304202-25GEA 62.5 EUR
APOSOR304202-25G
5-Hydroxy-o-toluylsäure 98%
5-Hydroxy-o-toluylsäure
Formel: C₈H₈O₃
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 182…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09833184
CAS-Nummer: 578-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR