5-Brom-2-furaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromfurfural
OR30421-5G
APOSOR30421-5GEA
21.9
EUR
APOSOR30421-5G
5-Brom-2-furaldehyd
5-Brom-2-furaldehyd
Formel:
C₅H₃BrO₂ Molecular Weight: 174.98 g/mol Siedepunkt: 112 °C/16mmHg Schmelzpunkt: 82…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00159501 CAS-Nummer: 1899-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...