1,3-Acetonedicarboxylic acid anhydride 97%

Lieferant: Apollo Scientific

Synonyme: 2H-Pyran-2,4,6(3H,5H)-trione

OR30430-250MG
APOSOR30430-250MGEA 260 EUR
APOSOR30430-250MG
1,3-Acetonedicarboxylic acid anhydride 97%
1,3-Acetonedicarboxylic acid anhydride
Formel: C₅H₄O₄
Molecular Weight: 128,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07783094
CAS-Nummer: 10521-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR