3,6-Dichloro-4-hydroxypyridazine 95+%

Lieferant: Apollo Scientific

Synonyme: 3,6-Dichloro-4-hydroxy-1,2-diazine, 3,6-Dichloropyridazin-4-ol

OR304303-1G
APOSOR304303-1GEA 23.2 EUR
APOSOR304303-1G
3,6-Dichloro-4-hydroxypyridazine 95+%
3,6-Dichloro-4-hydroxypyridazine
Formel: C₄H₂Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD04971276
CAS-Nummer: 2779-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR