2-(Azetidin-2-yl)propan-2-ol hydrochloride
Lieferant: Apollo Scientific
OR304319-1G
APOSOR304319-1GEA
1710
EUR
APOSOR304319-1G
2-(Azetidin-2-yl)propan-2-ol hydrochloride
2-(Azetidin-2-yl)propan-2-ol hydrochloride
Formel:
C₆H₁₃NO·ClH Molecular Weight: 151,634 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24509563 CAS-Nummer: 1824166-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...