4-(Propan-2-yl)azetidin-2-one

Lieferant: Apollo Scientific

OR304320-1G
APOSOR304320-1GEA 700 EUR
APOSOR304320-1G
4-(Propan-2-yl)azetidin-2-one
4-(Propan-2-yl)azetidin-2-one
Formel: C₆H₁₁NO
Molecular Weight: 113,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06217522
CAS-Nummer: 7486-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR