2-(1-Methylazetidin-3-yl)ethan-1-amine

Lieferant: Apollo Scientific

OR304322-1G
APOSOR304322-1GEA 877 EUR
APOSOR304322-1G
2-(1-Methylazetidin-3-yl)ethan-1-amine
2-(1-Methylazetidin-3-yl)ethan-1-amine
Formel: C₆H₁₄N₂
Molecular Weight: 114,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26936204
CAS-Nummer: 1542210-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR