3-Ethynylazetidine hydrochloride

Lieferant: Apollo Scientific

OR304323-1G
APOSOR304323-1GEA 0 EUR
APOSOR304323-1G
3-Ethynylazetidine hydrochloride
3-Ethynylazetidine hydrochloride
Formel: C₅H₇N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18911000
CAS-Nummer: 1426424-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR