(2S)-3,3-Dimethylazetidine-2-carboxylic acid
Lieferant: Apollo Scientific
OR304331-1G
APOSOR304331-1GEA
1340
EUR
APOSOR304331-1G
(2S)-3,3-Dimethylazetidine-2-carboxylic acid
(2S)-3,3-Dimethylazetidine-2-carboxylic acid
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30146439 CAS-Nummer: 1860033-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...