3-(Propan-2-yl)azetidin-3-ol hydrochloride
Lieferant: Apollo Scientific
OR304332-1G
OR304332-5G
APOSOR304332-1GEA
0
EUR
APOSOR304332-1G
APOSOR304332-5G
3-(Propan-2-yl)azetidin-3-ol hydrochloride
3-(Propan-2-yl)azetidin-3-ol hydrochloride
Formel:
C₆H₁₃NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD27959490 CAS-Nummer: 848192-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...