tert-Butyl 3-ethynyl-3-methylazetidine-1-carboxylate

Lieferant: Apollo Scientific

OR304334-1G
APOSOR304334-1GEA 3110 EUR
APOSOR304334-1G
tert-Butyl 3-ethynyl-3-methylazetidine-1-carboxylate
tert-Butyl 3-ethynyl-3-methylazetidine-1-carboxylate
Formel: C₁₁H₁₇NO₂
Molecular Weight: 195,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566173
CAS-Nummer: 1363381-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR