(2S)-2-(Methoxymethyl)azetidine
Lieferant: Apollo Scientific
OR304335-1G
APOSOR304335-1GEA
923
EUR
APOSOR304335-1G
(2S)-2-(Methoxymethyl)azetidine
(2S)-2-(Methoxymethyl)azetidine
Formel:
C₅H₁₁NO Molecular Weight: 101,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11977228 CAS-Nummer: 935668-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...