2-[(2S)-Azetidin-2-yl]propan-2-ol hydrochloride

Lieferant: Apollo Scientific

OR304337-1G
APOSOR304337-1GEA 0 EUR
APOSOR304337-1G
2-[(2S)-Azetidin-2-yl]propan-2-ol hydrochloride
2-[(2S)-Azetidin-2-yl]propan-2-ol hydrochloride
Formel: C₆H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31617919
CAS-Nummer: 935669-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR