2-[(2R)-Azetidin-2-yl]propan-2-ol hydrochloride

Lieferant: Apollo Scientific

OR304338-1G
APOSOR304338-1GEA 1560 EUR
APOSOR304338-1G
2-[(2R)-Azetidin-2-yl]propan-2-ol hydrochloride
2-[(2R)-Azetidin-2-yl]propan-2-ol hydrochloride
Formel: C₆H₁₃NO·ClH
Molecular Weight: 151,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31617920
CAS-Nummer: 2173637-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR