cis-3-(Propan-2-yloxy)cyclobutan-1-amine hydrochloride
Lieferant: Apollo Scientific
OR304342-1G
APOSOR304342-1GEA
923
EUR
APOSOR304342-1G
cis-3-(Propan-2-yloxy)cyclobutan-1-amine hydrochloride
cis-3-(Propan-2-yloxy)cyclobutan-1-amine hydrochloride
Formel:
C₇H₁₅NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD28892829 CAS-Nummer: 1909294-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...