1-(3-Hydroxycyclobutyl)ethan-1-one

Lieferant: Apollo Scientific

OR304344-1G
APOSOR304344-1GEA 1710 EUR
APOSOR304344-1G
1-(3-Hydroxycyclobutyl)ethan-1-one
1-(3-Hydroxycyclobutyl)ethan-1-one
Formel: C₆H₁₀O₂
Molecular Weight: 114,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24701736
CAS-Nummer: 30494-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR