3,3-Dimethylcyclobutan-1-amine hydrochloride

Lieferant: Apollo Scientific

OR304345-1G
APOSOR304345-1GEA 654 EUR
APOSOR304345-1G
3,3-Dimethylcyclobutan-1-amine hydrochloride
3,3-Dimethylcyclobutan-1-amine hydrochloride
Formel: C₆H₁₃N·ClH
Molecular Weight: 135,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22987614
CAS-Nummer: 1284247-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR