3-Amino-3-methylcyclobutan-1-one hydrochloride

Lieferant: Apollo Scientific

OR304350-1G
APOSOR304350-1GEA 1710 EUR
APOSOR304350-1G
3-Amino-3-methylcyclobutan-1-one hydrochloride
3-Amino-3-methylcyclobutan-1-one hydrochloride
Formel: C₅H₉NO·ClH
Molecular Weight: 135,592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30724194
CAS-Nummer: 2089255-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR