3-tert-Butylcyclobutanone

Lieferant: Apollo Scientific

OR304353-1G
APOSOR304353-1GEA 1560 EUR
APOSOR304353-1G
3-tert-Butylcyclobutanone
3-tert-Butylcyclobutanone
Formel: C₈H₁₄O
Molecular Weight: 126.196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19441103
CAS-Nummer: 20614-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR