2,2-Dimethylcyclobutan-1-one

Lieferant: Apollo Scientific

OR304354-1G
APOSOR304354-1GEA 1930 EUR
APOSOR304354-1G
2,2-Dimethylcyclobutan-1-one
2,2-Dimethylcyclobutan-1-one
Formel: C₆H₁₀O
Molecular Weight: 98,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22049843
CAS-Nummer: 1192-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR