Cyclobutane-1,3-diol
Lieferant: Apollo Scientific
OR304355-1G
APOSOR304355-1GEA
1150
EUR
APOSOR304355-1G
Cyclobutane-1,3-diol
Cyclobutane-1,3-diol
Formel:
C₄H₈O₂ Molecular Weight: 88,105 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22627868 CAS-Nummer: 63518-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...