3-(Hydroxymethyl)cyclobutan-1-ol

Lieferant: Apollo Scientific

OR304356-1G
APOSOR304356-1GEA 299 EUR
APOSOR304356-1G
3-(Hydroxymethyl)cyclobutan-1-ol
3-(Hydroxymethyl)cyclobutan-1-ol
Formel: C₅H₁₀O₂
Molecular Weight: 102,132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09743755
CAS-Nummer: 1245647-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR