N1,N1-Dimethylcyclobutane-1,3-diamine dihydrochloride

Lieferant: Apollo Scientific

OR304357-1G
APOSOR304357-1GEA 811 EUR
APOSOR304357-1G
N1,N1-Dimethylcyclobutane-1,3-diamine dihydrochloride
N1,N1-Dimethylcyclobutane-1,3-diamine dihydrochloride
Formel: C₆H₁₄N₂·2ClH
Molecular Weight: 187,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16621834
CAS-Nummer: 1229625-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR