3,3-Dimethylcyclobutan-1-ol
Lieferant: Apollo Scientific
OR304359-1G
APOSOR304359-1GEA
1340
EUR
APOSOR304359-1G
3,3-Dimethylcyclobutan-1-ol
3,3-Dimethylcyclobutan-1-ol
Formel:
C₆H₁₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11878436 CAS-Nummer: 54166-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...