3,3-Dimethylcyclobutan-1-one

Lieferant: Apollo Scientific

OR304364-1G
APOSOR304364-1GEA 1510 EUR
APOSOR304364-1G
3,3-Dimethylcyclobutan-1-one
3,3-Dimethylcyclobutan-1-one
Formel: C₆H₁₀O
Molecular Weight: 98.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22054450
CAS-Nummer: 1192-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR