cis-3-Methylcyclobutan-1-amine hydrochloride

Lieferant: Apollo Scientific

OR304367-1G
APOSOR304367-1GEA 1130 EUR
APOSOR304367-1G
cis-3-Methylcyclobutan-1-amine hydrochloride
cis-3-Methylcyclobutan-1-amine hydrochloride
Formel: C₅H₁₁N·ClH
Molecular Weight: 121,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12925785
CAS-Nummer: 1093951-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR