Methyl 2-(3-hydroxycyclobutyl)acetate
Lieferant: Apollo Scientific
OR304368-1G
APOSOR304368-1GEA
713
EUR
APOSOR304368-1G
Methyl 2-(3-hydroxycyclobutyl)acetate
Methyl 2-(3-hydroxycyclobutyl)acetate
Formel:
C₇H₁₂O₃ Molecular Weight: 144,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20134007 CAS-Nummer: 1148130-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...