Methyl trans-2-aminocyclobutanecarboxylate hydrochloride

Lieferant: Apollo Scientific

OR304370-1G
APOSOR304370-1GEA 1930 EUR
APOSOR304370-1G
Methyl trans-2-aminocyclobutanecarboxylate hydrochloride
Methyl trans-2-aminocyclobutanecarboxylate hydrochloride
Formel: C₆H₁₁NO₂·ClH
Molecular Weight: 165.618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30490769
CAS-Nummer: 1159279-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR