3-(3-Oxocyclobutyl)propanoic acid
Lieferant: Apollo Scientific
OR304372-1G
APOSOR304372-1GEA
877
EUR
APOSOR304372-1G
3-(3-Oxocyclobutyl)propanoic acid
3-(3-Oxocyclobutyl)propanoic acid
Formel:
C₇H₁₀O₃ Molecular Weight: 142,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24503127 CAS-Nummer: 1380291-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...