3-(Benzyloxy)-2,2-dimethylcyclobutan-1-one

Lieferant: Apollo Scientific

OR304375-1G
APOSOR304375-1GEA 1150 EUR
APOSOR304375-1G
3-(Benzyloxy)-2,2-dimethylcyclobutan-1-one
3-(Benzyloxy)-2,2-dimethylcyclobutan-1-one
Formel: C₁₃H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD21755679
CAS-Nummer: 2063-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR